Electrical laboratory jacks, Lift 240
Supplier: BOCHEM
11220
BOCH11220EA 7530 SGD
BOCH11220
Electrical laboratory jacks, Lift 240
Laboratory Jacks
Stainless steel ¹⁸/₁₀. Remote control operated lab jack with integrated motor for use in hoods.
  • Chemically resistant PTFE bellows protect the electronic components
  • Stable stand and wobble-free lifting and lowering
  • The end stop can be manually adjusted from the outside
Max. load capacity (dynamic): 25 kg