Transport trolleys
Supplier: BOCHEM
11418 11417
BOCH11418EA 4170 SGD
BOCH11418 BOCH11417
Transport trolleys
Trolleys
These transport trolleys are made of stainless steel with two guide and two fixed rolls.
  • Surface matt finish
  • Robust pushing handle
Load capacity: 300 kg
Order Now
SPECIFICATIONS
Environmentally Preferable