Laboratory tables, with sliding door
Supplier: BOCHEM
11383 11380 11382 11384 11391 11392 11393 11394 11402 11403 11381 11583 11580 11404 11582 11584 11591 11592 11581 11791 11792 11793 11794 11991 11992 11993 11994 11604 11593 11594 11603 11602
BOCH11383EA 18720 SGD
BOCH11383 BOCH11380 BOCH11382 BOCH11384 BOCH11391 BOCH11392 BOCH11393 BOCH11394 BOCH11402 BOCH11403 BOCH11381 BOCH11583 BOCH11580 BOCH11404 BOCH11582 BOCH11584 BOCH11591 BOCH11592 BOCH11581 BOCH11791 BOCH11792 BOCH11793 BOCH11794 BOCH11991 BOCH11992 BOCH11993 BOCH11994 BOCH11604 BOCH11593 BOCH11594 BOCH11603 BOCH11602
Laboratory tables, with sliding door
Furniture Tables and Workbenches
These laboratory tables are made up of 18/10 stainless steel and height adjustable.
  • Drawer face with user friendly handle bar
  • Noise protection
Working plate with matt finishing and corners are continuously welded and polished.
Drawer with guard rails made of stainless steel to mount GN-drawer. Drawers can be taken off for cleaning, but are secured against drawing out.
Order Now
SPECIFICATIONS
Environmentally Preferable