Dissection set
Supplier: BOCHEM
12017
BOCH12017EA 121 SGD
BOCH12017
Dissection set
Dissection Sets
Stainless steel. This dissection set is designed for academic applications. Includes.
  • One pair of scissors with two sharp cutting tips
  • One pair of scissors with a sharp and a blunt cutting tip
  • Two scalpels
  • Two forceps
  • Two dissection needles
Order Now
SPECIFICATIONS
Environmentally Preferable