Cork borers
Supplier: BOCHEM
12435 12431 12433
BOCH12435EA 1940 SGD
BOCH12435 BOCH12431 BOCH12433
Cork borers
Samplers Solid Samplers
Cork borers made of hard coated nickel-plated brass.
Delivery information: Include a cleaning needle.
Order Now
SPECIFICATIONS
Environmentally Preferable