Weighing forceps
Supplier: BOCHEM
1740
BOCH1740EA 27.7 SGD
BOCH1740
Weighing forceps
Forceps
Stainless steel 18/10, antimagnetic, with plastic tips.
  • Ideal for picking up and handling precision weights
  • Max. temperature 145 °C