Sink table, with ground plate
Supplier: BOCHEM
21805 21806
BOCH21805EA 12020 SGD
BOCH21805 BOCH21806
Sink table, with ground plate
Furniture Casework
Sink table with place to keep two sinks 400×400×200 mm, with drain 38 mm.
Order Now
SPECIFICATIONS
Environmentally Preferable