Beaker tongs, stainless steel
Supplier: BOCHEM
2220
BOCH2220EA 54 SGD
BOCH2220
Beaker tongs, stainless steel
Tongs
Stainless steel ¹⁸/₁₀, antimagnetic and electrolytical polished.
  • Jaws are insulated with ceramic
  • For beakers 100 to 1500 ml
Order Now
SPECIFICATIONS
Environmentally Preferable