Double spatulas, micro
Supplier: BOCHEM
3017 3024 3023 3011 3030 3019 3010 3012 3016 3018 3020 3022 3025 3026 3027 3028 3029 3013 3015 30245 30235 30225 30145 30135 3021 3014
BOCH3017EA 6 SGD
BOCH3017 BOCH3024 BOCH3023 BOCH3011 BOCH3030 BOCH3019 BOCH3010 BOCH3012 BOCH3016 BOCH3018 BOCH3020 BOCH3022 BOCH3025 BOCH3026 BOCH3027 BOCH3028 BOCH3029 BOCH3013 BOCH3015 BOCH30245 BOCH30235 BOCH30225 BOCH30145 BOCH30135 BOCH3021 BOCH3014
Double spatulas, micro
Spatulas
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
  • With round handle, one flat and one rounded end
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
Order Now
SPECIFICATIONS
Environmentally Preferable