Micro double spatulas
Supplier: BOCHEM
3053 3047 3046 3057
BOCH3053EA 104 SGD
BOCH3053 BOCH3047 BOCH3046 BOCH3057
Micro double spatulas
Spatulas
Nickel. Multipurpose spatulas for a wide range of uses.
  • With round handle, one flat and one rounded end
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance