Double spatulas, bent
Supplier: BOCHEM
3100G 3101G 3103G 3104G 3105G
BOCH3100GEA 7 SGD
BOCH3100G BOCH3101G BOCH3103G BOCH3104G BOCH3105G
Double spatulas, bent
Spatulas
Stainless steel 18/10, anti-magnetic. Multi-purpose spatula for a wide range of uses.
  • With round handle, one flat and one rounded end
  • Curved shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
Order Now
SPECIFICATIONS
Environmentally Preferable