Double spatulas
Supplier: BOCHEM
3100 3102 3103 3105 3107 3104 3108 31005 31015 31025 31035 31055 31045 3101
BOCH3100EA 5.6 SGD
BOCH3100 BOCH3102 BOCH3103 BOCH3105 BOCH3107 BOCH3104 BOCH3108 BOCH31005 BOCH31015 BOCH31025 BOCH31035 BOCH31055 BOCH31045 BOCH3101
Double spatulas
Spatulas
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
  • Round handle, one flat and one rounded end
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance