Double spatulas, micro
Supplier: BOCHEM
3300 3302 3303 3304 3301
BOCH3300EA 6.8 SGD
BOCH3300 BOCH3302 BOCH3303 BOCH3304 BOCH3301
Double spatulas, micro
Spatulas
Stainless steel 18/10, anti-magnetic. With round handle, one flat and one scoop shaped, rounded spatula end.
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
Order Now
SPECIFICATIONS
Environmentally Preferable