Pharmacist's spatulas
Supplier: BOCHEM
3500 3501 3502 3503 3504
BOCH3500EA 13.6 SGD
BOCH3500 BOCH3501 BOCH3502 BOCH3503 BOCH3504
Pharmacist's spatulas
Spatulas
These spatulas feature a stainless steel blade and a PVC handle.
  • Suitable for dishwasher
  • Autoclavable