Vibration spatula
Supplier: BOCHEM
3540
BOCH3540EA 48.1 SGD
BOCH3540
Vibration spatula
Spatulas
Stainless steel, plastic shaft. A toothed wheel is turned by a finger making the spatula blade vibrate. This action allows for accurate adding of material.
Order Now
SPECIFICATIONS
Environmentally Preferable