Double spatulas, coated
Supplier: BOCHEM
3700 3701 3702
BOCH3700EA 70.4 SGD
BOCH3700 BOCH3701 BOCH3702
Double spatulas, coated
Spatulas
Double-ended spatulas made of stainless steel with PTFE coating.
Order Now
SPECIFICATIONS
Environmentally Preferable