Measuring spoons, set
Supplier: BOCHEM
3801
BOCH3801EA 32 SGD
BOCH3801
Measuring spoons, set
Spoons
Stainless steel 18/10. Practical and convenient set of four measuring spoons.
  • Heat and chemical resistant
  • Ringed set keeps all sizes together
Delivery information: Each set includes 1,25 ml, 2,5 ml, 5 ml and 15 ml spoons.
Order Now
SPECIFICATIONS
Environmentally Preferable