Retort clamp, four pronged
Supplier: BOCHEM
5423
BOCH5423EA 413 SGD
BOCH5423
Retort clamp, four pronged
Clamps General Purpose Clamps
This four pronged retort clamp is made of stainless steel.
  • Fingers are coated with silicone to protect clamped items