Burette clamps, double
Supplier: BOCHEM
5600
BOCH5600EA 170 SGD
BOCH5600
Burette clamps, double
Clamps General Purpose Clamps
Aluminium.
  • With bosshead for fixing to retort stand with Ø 0 to 13
  • With PVC rolls
  • Distance between rolls 170 mm
Order Now
SPECIFICATIONS
Environmentally Preferable