Burette and condenser clamps, with swivelling bosshead
Supplier: BOCHEM
5630
BOCH5630EA 88.7 SGD
BOCH5630
Burette and condenser clamps, with swivelling bosshead
Clamps General Purpose Clamps
Burette clamps made of chromium plated steel.
  • Fingers are coated with PVC to protect clamped items
  • With swivelling bosshead (16,5 mm)
  • Angle: 0 to 360°