Support clips
Supplier: BOCHEM
580 582 583 584 581 590 591 592 593 594 595 596 597 598 599 600
BOCH580EA 93.5 SGD
BOCH580 BOCH582 BOCH583 BOCH584 BOCH581 BOCH590 BOCH591 BOCH592 BOCH593 BOCH594 BOCH595 BOCH596 BOCH597 BOCH598 BOCH599 BOCH600
Support clips
Retort Rings
These support clips are coated with ceramic inside.
  • For tubes and cylindrical vessels
  • With fitting
Ordering information: The shafts (rods) need to be ordered separately.