Bunsen burners, with stopcock
Supplier: BOCHEM
7360
BOCH7360EA 435 SGD
BOCH7360
Bunsen burners, with stopcock
Heaters Gas Burners
This multigas burner can reach a maximum temperature of 1300 °C and has an anti-slip rubber coating under the base.
  • With air regulation
Order Now
SPECIFICATIONS
Environmentally Preferable