Evaporating dishes with spout
Supplier: BOCHEM
8023 8020 8021 8022 8024
BOCH8023EA 206 SGD
BOCH8023 BOCH8020 BOCH8021 BOCH8022 BOCH8024
Evaporating dishes with spout
Trays/Dishes/Pots Evaporating Basins/Dishes
Evaporating dish, made of nickel, with spout.
  • Corrosion resistant
  • Very good thermal resistance
  • Smooth surfaces for easy cleaning
Multipurpose tray ideal for a wide variety of uses.
Order Now
SPECIFICATIONS
Environmentally Preferable