Laboratory trays, with rim
Supplier: BOCHEM
8454 8455 8456 8457 8459 8399 8395 8396 8397 8398
BOCH8454EA 64.3 SGD
BOCH8454 BOCH8455 BOCH8456 BOCH8457 BOCH8459 BOCH8399 BOCH8395 BOCH8396 BOCH8397 BOCH8398
Laboratory trays, with rim
Trays/Dishes/Pots Evaporating Basins/Dishes
Stainless steel, ¹⁸/₁₀.
  • Corrosion resistant
  • Excellent thermal resistance
  • Smooth surfaces for easy cleaning
Multipurpose tray ideal for a wide variety of uses.
Order Now
SPECIFICATIONS
Environmentally Preferable