Instrument trays, with lid
Supplier: BOCHEM
8400 8404 8406 8407
BOCH8400EA 181 SGD
BOCH8400 BOCH8404 BOCH8406 BOCH8407
Instrument trays, with lid
Trays/Dishes/Pots Instrument Trays/Dishes
Stainless steel, corrosion-resistant, 18/10.
  • Special corner shape for fast, easy cleaning
  • Polished electrolytically, anti-magnetic
  • Good chemical resistance, very good thermal resistance
With a cover that enables the instrument dishes to be stacked.
Order Now
SPECIFICATIONS
Environmentally Preferable