Laboratory pots, with lid
Supplier: BOCHEM
8566 8567 8568
BOCH8566EA 84.5 SGD
BOCH8566 BOCH8567 BOCH8568
Laboratory pots, with lid
Trays/Dishes/Pots Pots
Aluminium.
  • Fast, easy cleaning
  • Good chemical resistance
  • Very good thermal resistance
Multipurpose pots for a wide range of uses.
Order Now
SPECIFICATIONS
Environmentally Preferable