Laboratory bowls
Supplier: BOCHEM
8600 8601 8602 8603 8605 8606 8607 8608 8609 8610 8611
BOCH8600EA 38 SGD
BOCH8600 BOCH8601 BOCH8602 BOCH8603 BOCH8605 BOCH8606 BOCH8607 BOCH8608 BOCH8609 BOCH8610 BOCH8611
Laboratory bowls
Trays/Dishes/Pots Laboratory Trays/Dishes
Bowls are high form, and made of stainless steel 18/10.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean
Round multipurpose dishes for a wide range of uses.
Order Now
SPECIFICATIONS
Environmentally Preferable