Pharmacy scoops
Supplier: BOCHEM
8810 8811 8812 8813 8809
BOCH8810EA 334 SGD
BOCH8810 BOCH8811 BOCH8812 BOCH8813 BOCH8809
Pharmacy scoops
Scoops
These chemical scoops made from stainless steel 18/10 are ideal for use in pharmaceutical and food industries.
  • With welded handle
  • Food contact approved
Order Now
SPECIFICATIONS
Environmentally Preferable