Pharma scoops
Supplier: BOCHEM
8838 8837 8836 8835
BOCH8838EA 146 SGD
BOCH8838 BOCH8837 BOCH8836 BOCH8835
Pharma scoops
Scoops
Stainless steel. These scoops are ideal for sample taking in the pharmaceutical and food industries.
  • With hanging loop
  • Seamless fixed handle
Order Now
SPECIFICATIONS
Environmentally Preferable