Sterilising box for petri dishes
Supplier: BOCHEM
8961
BOCH8961EA 592 SGD
BOCH8961
Sterilising box for petri dishes
Racks Petri Dish Racks
Sterilising box made of stainless steel, for up to ten Petri dishes with Ø 100 mm.