Overhead stirrer tool, radial stirrer
Supplier: BOCHEM
9030
BOCH9030EA 337 SGD
BOCH9030
Overhead stirrer tool, radial stirrer
Stirrers Stirring Tools
Radial stirrer made of stainless steel.
  • Ideal for stirring liquids at middle speed range
  • Suitable for stirring low and medium viscous media
  • Creates radial flow
  • Ideal for aeration of liquids