Stirrers, propeller, with four paddles
Supplier: BOCHEM
9054 9053 9055 9056 9057 9058
BOCH9054EA 78.3 SGD
BOCH9054 BOCH9053 BOCH9055 BOCH9056 BOCH9057 BOCH9058
Stirrers, propeller, with four paddles
Stirrers Stirring Tools
Propeller stirrer made of stainless steel with four vertical paddles.
  • Mixing of media with low and middle viscosity
  • Radial flow