Overhead stirrer tools, paddle stirrers
Supplier: BOCHEM
9406 9408 9407 9403 9404 9405
BOCH9406EA 85.2 SGD
BOCH9406 BOCH9408 BOCH9407 BOCH9403 BOCH9404 BOCH9405
Overhead stirrer tools, paddle stirrers
Stirrers Stirring Tools
Paddle stirrers made of stainless steel, with three holes.
  • Ideal for stirring liquids at middle speed range
  • Suitable for stirring low to medium viscous media
  • Creates radial flow