Overhead stirrer tools, paddle stirrers
Supplier: BOCHEM
9503 9507 9504 9505 9506 9508
BOCH9503EA 81.6 SGD
BOCH9503 BOCH9507 BOCH9504 BOCH9505 BOCH9506 BOCH9508
Overhead stirrer tools, paddle stirrers
Stirrers Stirring Tools
Paddle stirrers made of stainless steel, with six holes.
  • Ideal for stirring liquids at middle speed range
  • Suitable for stirring low to medium viscous media
  • Creates radial flow