Stirrers, propeller, with two paddles
Supplier: BOCHEM
9704 9705 9703
BOCH9704EA 64.9 SGD
BOCH9704 BOCH9705 BOCH9703
Stirrers, propeller, with two paddles
Stirrers Stirring Tools
Propeller stirrer made of stainless steel with two fixed paddles.
  • Mixing of media with low and middle viscosity
  • Radial flow