Electronic overhead stirrer, RS9001
Supplier: BOCHEM
RS9001
BOCHRS9001EA 4800 SGD
BOCHRS9001
Electronic overhead stirrer, RS9001
Stirrers Overhead Stirrers
Stirrer with gear for stirring media with high viscosity. Laboratory stirrer with variable and constant speed range for media.
  • Digital LCD display
  • Digital timer function (99 hours)
  • Speed range infinitely variable at/with high quiet running
  • Convenient user interface
  • Overload protection and over-heat protection functions
  • Smooth start and change of the stirring speed
  • Impeller can be plugged through
Suitable for most stirring tasks in a laboratory.