Hybridisation apparatus, Big Shot III™ and Little Shot III™
Supplier: BOEKEL SCIENTIFIC
230402-EU 230402-UK
BOEI230402-EUEA 0 SGD
BOEI230402-EU BOEI230402-UK
Hybridisation apparatus, Big Shot III™ and Little Shot III™
Ovens Hybridisation Ovens
Hybridisation and wash methods vary according to application and personal choice. Boekel Scientific offers a range of hybridisation incubators with features that include digital display for ease and reproducibility of temperature setting; microprocessor or proportional temperature control; rapid heat-up, optimised heater power and natural or forced air convection.
  • Forced air convection, coupled with PID temperature control, combine to provide rapid heat up and excellent temperature stability
  • Unit is stackable and provided with a reversible door to optimise
More Product Information