Goggles, Cobra
Supplier: Bollé Safety
COBFSPSI
BOLECOBFSPSIEA 38.5 SGD
BOLECOBFSPSI
Goggles, Cobra
Goggles
Low profile goggles with soft foam seal, adjustable strap and non slip bridge. This version of the Cobra combines the comfort of an eyeshield with the fit and seal of an all-purpose goggle.
  • Clear anti-static PC lenses for 180° panoramic field of view
  • Interchangeable comfort foam
  • Lenses with anti-fog and anti-scratch coating for improved resistance
Weight: 26 g
Certifications: EN 166, EN 170