Safety spectacles, Cobra HD
Supplier: Bollé Safety
COBHDPI
BOLECOBHDPIEA 30.8 SGD
BOLECOBHDPI
Safety spectacles, Cobra HD
Safety Glasses
Cobra spectacles are multipurpose eyewear with a panoramic visual field. The side arms can be exchanged by a head band.
  • Non-slip bridge and temples
  • Clear, coated PC lens with anti-fog and anti-scratch properties for 180° view
  • HD lens with the highest level of optical clarity and aquaphobic properties
Weight: 25 g
Weight: 25 g
Certifications: EN 166, EN 170
Ordering information: The Cobra spectacles are also available with a foam inner layer and head band (rather than arms), which allows them to be rated as a goggle for protection against liquid droplets.
Order Now
SPECIFICATIONS
Environmentally Preferable