Safety spectacles, Cobra
Supplier: Bollé Safety
COBPSI
BOLECOBPSIEA 28.6 SGD
BOLECOBPSI
Safety spectacles, Cobra
Safety Glasses
Cobra spectacles are esential all rounder with a 180° view panoramic visual field. Available as temple or strap and foam version with interchangeable foam. The option of swapping the temples for an adjustable strap and foam seal means COBRA is a highly versatile model.
  • Non-slip bridge and temples
  • Clear, coated PC lens with anti-fog and anti-scratch properties
  • Pivoting temples
Weight: 25 g
Certifications: EN 166, EN 170