Goggles, coverall
Supplier: Bollé Safety
COVERSI
BOLECOVERSIEA 28.4 SGD
BOLECOVERSI
Goggles, coverall
Goggles
Comfortable goggles with adjustable elastic straps.
  • Fit comfortably over most prescription eyewear
  • High flow ventilation with four indirect ventilators
  • Resistant to molten metal and hot solids, dust and liquid aerosols
Certifications: EN 166