Safety spectacles, Silium
Supplier: Bollé Safety
SILPSI SILPSF
BOLESILPSIEA 34.2 SGD
BOLESILPSI BOLESILPSF
Safety spectacles, Silium
Safety Glasses
These slim and attractively designed spectacles feature a metal, nickel-free frame and a curved lens.
  • Flexible temples bend up to 160%, equipped with comfort pads
  • Flexible fittings in the nose bridge offer extra comfort
Weight: 34 g
EN 166, EN 170
Certifications: EN 166, EN 170