Slam Safety Spectacles
Supplier: Bollé Safety
SLAPSI
BOLESLAPSIEA 17.1 SGD
BOLESLAPSI
Slam Safety Spectacles
Safety Glasses
Modern design spectacles with extra-easy fitting and comfort.
  • Panoramic visual field
  • Anti-scratch, anti-fog, anti-static coatings
  • Ultra enveloping
  • Comfort temples
Weight: 23 g.
Certifications: EN 166, EN 170 (clear lens), EN 172 (smoke lens).
Delivery information: Supplied with adjustable cord.