Safety spectacles, Solis
Supplier: Bollé Safety
SOLIFLASH
BOLESOLIFLASHEA 0 SGD
BOLESOLIFLASH
Safety spectacles, Solis
Safety Glasses
Solis spectacles offer a wrap-around style with the benefits of development of modern materials.
  • Non slip TIPGRIP temples
  • Non slip bridge
  • Integral side shields
  • Polarised coating
Weight: 25 g
Certifications: EN 166, EN 172
Delivery information: Supplied with microfibre pouch.