Cover spectacles, Squale
Supplier: Bollé Safety
SQUPSI
BOLESQUPSIEA 19.1 SGD
BOLESQUPSI
Cover spectacles, Squale
Safety Glasses
Cover glasses (OTG) give excellent protection for wearers of prescription eyewear. Supplied with adjustable cord.
  • Adjustable Nylon temples
  • Moulded lens, brow guard and side shields
  • Indirect ventilation
Weight: 43 g
Certifications: EN 166, EN 170