Ultrasonic baths, Bransonic™ B200
Supplier: BRANSON
000-001-100
BRAN000-001-100EA 472 SGD
BRAN000-001-100
Ultrasonic baths, Bransonic™ B200
Ultrasonic Baths
The B200 ultrasonic bath can clean smaller objects such as geological samples, electrical/electronic components and many small instruments. Suitable for cleaning contact lenses, spectacles and other optical articles. Effective for removing surface contamination such as fingerprints and dust.
  • A versatile bath that can be used anywhere
  • Compact design
  • Stainless steel container with 0,5 L capacity contained within a tough plastic housing
  • With built-in 5 minute timer that switches the bath off automatically
Delivery information: Lid and parts basket areMore Product Information