Universal multi-pole breaker lockout
Supplier: Brady
66321
BRDY66321EA 44.5 SGD
BRDY66321
Universal multi-pole breaker lockout
Security Locks
This universal breaker lockout is compatible with most two or three pole breakers with a tie-bar, such as GE ITE, Sylvania, Challenger, Bryant, Westinghouse and General Switch Company.
  • Made of moulded glass-filled nylon
Use thumbscrews to clamp lockout securely over tie-bar, then insert lock to prevent clamp from being loosened.
Max. shackle diameter: 7,00 mm
Min. shackle clearance: 20,00 mm