Non-return valve
Supplier: Brand
134002
BRND134002EA 11.2 SGD
BRND134002
Non-return valve
Valves Non Return Valves
HDPE, transparent. Prevents backflow. With FKM valve disc not suitable for pressure applications.
  • Good chemical resistance
  • Good thermal resistance
  • With tapered ends to ensure that the tube is held firmly