Volumetric flasks, class A, white markings, BLAUBRAND®
Supplier: Brand
2.37407
BRND2.37407EA 159 SGD
BRND2.37407
Volumetric flasks, class A, white markings, BLAUBRAND®
Flasks Volumetric Flasks
Borosilicate glass 3.3, amber, with PP stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast white enamel
  • DE-M marking
  • Total order from one batch, supplied with batch certificate
*Wide neck.
Certifications: DIN EN ISO 1042