Pipette tray
Supplier: Brand
28300
BRND28300EA 76.7 SGD
BRND28300
Pipette tray
Trays/Dishes/Pots Pipette Trays
Made of white PVC.
  • Good temperature resistance
  • Good chemical resistance
  • Easy to clean
Practical tray for pipettes of any size.